missing translation for 'onlineSavingsMsg'
Learn More

Isopentylamine, 99%

Product Code. 15498799
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15498799 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15498799 Supplier Thermo Scientific Alfa Aesar Supplier No. L02260.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-85-7 | C5H13N | 87.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 107-85-7
Melting Point -60°C
Density 0.751
pH 12.1
Boiling Point 95°C to 97°C
Flash Point −1°C (30°F)
Odor Amine-like
Molecular Formula C5H13N
Refractive Index 1.408
MDL Number MFCD00008203
Quantity 25 g
UN Number UN1106
Beilstein 1209230
Sensitivity Air Sensitive
Merck Index 14,5112
Synonym isoamylamine, isopentylamine, 3-methylbutylamine, 1-amino-3-methylbutane, monoisoamylamine, leucamine, 1-butanamine, 3-methyl, 3-methylbutanamine, isovalerylamine, monoisopentylamine
InChI Key BMFVGAAISNGQNM-UHFFFAOYSA-N
SMILES CC(C)CCN
IUPAC Name 3-methylbutan-1-amine
Molecular Weight (g/mol) 87.166
PubChem CID 7894
ChEBI CHEBI:43689
Formula Weight 87.17
Percent Purity 99%
Chemical Name or Material Isopentylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isopentylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Isoamylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.