missing translation for 'onlineSavingsMsg'
Learn More

Isophorone diisocyanate, mixture of isomers, 98%

Product Code. 11333227
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333227 500 g 500g
11313227 5 g 5g
11323227 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11333227 Supplier Thermo Scientific Alfa Aesar Supplier No. L13759.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 4098-71-9 | C12H18N2O2 | 222.29 g/mol

Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isophorone diisocyanate is useful for the preparation of polyurethane plastics. It is also used to make polyurethane coatings and in enamel coatings, which are resistant to abrasion and degradation from ultraviolet light. Further, it is used as coatings for automotive, flooring and roofing as well as paint applied to aircraft.

Solubility
Miscible with esters, ketones, ethers, and aromatic and aliphatic hydrocarbons.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4098-71-9
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.29
MDL Number MFCD00064956
InChI Key NIMLQBUJDJZYEJ-UHFFFAOYNA-N
Synonym isophorone diisocyanate, ipdi, isophorone diamine diisocyanate, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethylcyclohexane, 3-isocyanatomethyl-3,5,5-trimethylcyclohexylisocyanate, cyclohexane, 5-isocyanato-1-isocyanatomethyl-1,3,3-trimethyl, 3-isocyanatomethyl-3,5,5-trimethylcyclohexyl isocyanate, ccris 6252, isocyanic acid, methylene 3,5,5-trimethyl-3,1-cyclohexylene ester, isophorone diisocyanate diisocyanates
PubChem CID 169132
ChEBI CHEBI:53214
IUPAC Name 5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
SMILES CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O

Specifications

Melting Point -60°C
Density 1.061
Boiling Point 153°C (10 mmHg)
Flash Point 163°C (325°F)
Refractive Index 1.484
Quantity 500 g
UN Number 2290
Beilstein 2726467
Sensitivity Moisture sensitive
Solubility Information Miscible with esters,ketones,ethers,and aromatic and aliphatic hydrocarbons.
Formula Weight 222.29
Percent Purity 98%
Chemical Name or Material Isophorone Diisocyanate, mixture of isomers
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.