missing translation for 'onlineSavingsMsg'
Learn More

Isophorone Diisocyanate, TRC

Product Code. 30009905
Change view
Click to view available options
Quantity:
10 mL
50 mL
100 mL
Unit Size:
100mL
10mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009905 50 mL 50mL
30004385 100 mL 100mL
18000950 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30009905 Supplier Toronto Research Chemicals Supplier No. TRCI82197850ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4098-71-9
Molecular Formula C12 H18 N2 O2
Molecular Weight (g/mol) 222.28
Synonym Isocyanic acid, methylene(3,5,5-trimethyl-3,1-cyclohexylene) ester (7CI,8CI), 5-Isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane, 1,3,3-Trimethyl-1-(isocyanatomethyl)-5-isocyanatocyclohexane, 1-(Isocyanatomethyl)-5-isocyanato-1,3,3-trimethylcyclohexane, 1-Isocyanato-3,3,5-trimethyl-5-(isocyanatomethyl)cyclohexane, 1-Isocyanato-3-(isocyanatomethyl)-3,5,5-trimethylcyclohexane, 1-Isocyanato-5-(isocyanatomethyl)-3,3,5-trimethylcyclohexane, 3,3,5-Trimethyl-5-(isocyanatomethyl)cyclohexyl isocyanate, 3-(Isocyanatomethyl)-3,5,5-trimethylcyclohexyl isocyanate, IPDI, Isophorone diisocyanate, PDI, Z 4770, 5-Isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane, 1,3,3-Trimethyl-1-(isocyanatomethyl)-5-isocyanatocyclohexane, 1-(Isocyanatomethyl)-5-isocyanato-1,3,3-trimethylcyclohexane, 1-Isocyanato-3,3,5-trimethyl-5-(isocyanatomethyl)cyclohexane, 1-Isocyanato-3-(isocyanatomethyl)-3,5,5-trimethylcyclohexane, 1-Isocyanato-5-(isocyanatomethyl)-3,3,5-trimethylcyclohexane, 3,3,5-Trimethyl-5-(isocyanatomethyl)cyclohexyl isocyanate, 3-(Isocyanatomethyl)-3,5,5-trimethylcyclohexyl isocyanate, Carbitol acetate-isophorone Diisocyanate Copolymer, IPDI, PDI, Z 4770
IUPAC Name 5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
SMILES CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O

Specifications

Quantity 50 mL
InChI Formula InChI=1S/C12H18N2O2/c1-11(2)4-10(14-9-16)5-12(3,6-11)7-13-8-15/h10H,4-7H2,1-3H3
Formula Weight 222.1368
Chemical Name or Material Isophorone Diisocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.