missing translation for 'onlineSavingsMsg'
Learn More

Isophthalic Acid, TRC

Product Code. 30135947
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135947 500 mg 500mg
30133187 2.5 g 2.5g
17948524 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135947 Supplier Toronto Research Chemicals Supplier No. TRCI822008500MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-91-5
Molecular Formula C8 H6 O4
Molecular Weight (g/mol) 166.13
Synonym Isophthalic Acid, 1,3-Benzenedicarboxylic acid (9CI, ACI), Isophthalic acid (8CI), 3-Carboxybenzoic acid, HA 20, HA 20 (acid), Isoterephthalic acid, m-Benzenedicarboxylic acid, m-Carboxybenzoic acid, m-Dicarboxybenzene, m-Phthalic acid, m-Phthalic acid, NSC 15310
IUPAC Name benzene-1,3-dicarboxylic acid
SMILES OC(=O)c1cccc(c1)C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H6O4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4H,(H,9,10)(H,11,12)
Formula Weight 166.0266
Chemical Name or Material Isophthalic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.