missing translation for 'onlineSavingsMsg'
Learn More

Isophthalonitrile, 97+%

Product Code. 11436756
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11436756

Brand: Thermo Scientific Alfa Aesar B23900.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 626-17-5 | C8H4N2 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-17-5
Molecular Formula C8H4N2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00001795
InChI Key LAQPNDIUHRHNCV-UHFFFAOYSA-N
Synonym isophthalonitrile, 1,3-dicyanobenzene, isophthalodinitrile, 1,3-benzenedicarbonitrile, m-dicyanobenzene, 3-cyanobenzonitrile, m-benzenedinitrile, isoftalodinitril, 1,3-benzodinitrile, m-phthalodinitrile
PubChem CID 12276
ChEBI CHEBI:38218
IUPAC Name benzene-1,3-dicarbonitrile
SMILES N#CC1=CC(=CC=C1)C#N

Specifications

Melting Point 159°C to 163°C
Density 1.3
Boiling Point 288°C
Flash Point >150°C (302°F)
Odor Almond-like
Quantity 250 g
Beilstein 1072209
Formula Weight 128.13
Percent Purity 98%
Chemical Name or Material Isophthalonitrile
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.