missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoprene, 98%, stabilized
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
500mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -146.0°C |
| Color | Colorless to Yellow |
| Density | 0.6800g/mL |
| Boiling Point | 34.0°C |
| Flash Point | −48°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | H2C=CHC(CH3)=CH2 |
| Refractive Index | 1.4205 to 1.4225 |
| Show More |
Safety and Handling
Product Identifier
- Isoprene, stabilized
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 1B
- Flammable liquid Category 1
- Germ cell mutagenicity Category 2
- H224-Extremely flammable liquid and vapour.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Isoprene, stabilized
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction