missing translation for 'onlineSavingsMsg'
Learn More

Isoprene, 98%, stabilized

Product Code. 10448850 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
2.5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10448850 100 mL Glass bottle 100mL
10133550 500 mL Glass bottle 500mL
10265450 1 L Glass bottle 1 L
10193400 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10448850 Supplier Thermo Scientific Acros Supplier No. 122671000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78-79-5 | C5H8 | 68.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-79-5 , 98-29-3
Molecular Formula C5H8
Molecular Weight (g/mol) 68.11
MDL Number MFCD00008600
InChI Key RRHGJUQNOFWUDK-UHFFFAOYSA-N
Synonym isoprene, 2-methyl-1,3-butadiene, isopentadiene, 2-methylbutadiene, 1,3-butadiene, 2-methyl, 2-methyldivinyl, beta-methylbivinyl, isopreno, isoterpene, isopren
PubChem CID 6557
ChEBI CHEBI:35194
IUPAC Name 2-methylbuta-1,3-diene
SMILES CC(=C)C=C

Specifications

Melting Point -146.0°C
Color Colorless to Yellow
Density 0.6800g/mL
Boiling Point 34.0°C
Flash Point −48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHC(CH3)=CH2
Refractive Index 1.4205 to 1.4225
Quantity 100 mL
Beilstein 01, 252
Fieser 13,196
Merck Index 15, 5247
Specific Gravity 0.68
Solubility Information Solubility in water: 0.7g/L (25°C). Other solubilities: miscible with alcohol,ether and acetone,oils/fat
Stabilizer 100 to 250ppm 4-tert-butylcatechol
Viscosity 0.225 mPa.s (15°C)
Formula Weight 68.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isoprene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isoprene, stabilized
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Flammable liquid Category 1
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-methyl-1,3-Butadiene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.