missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, 99.8+%, for Electronic Use, Fisher Chemical™

Product Code. 12608655 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic Container
Unit Size:
2.5 L
25 L
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12608655 25 L Metal Can 25 L
10754191 2.5 L Amber Glass Bottle 2.5 L
10297233 5 L Plastic Container 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 12608655 Supplier Fisher Chemical Supplier No. P/7555/25
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number 11674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Spezifikation

pH 7
Melting Point -89.5°C
Color Colorless
Quantity 25 L
Packaging Metal Can
Physical Form Liquid
Vapor Pressure 43mmHg at 20°C
Viscosity 2.27 mPaS at 20°C
Boiling Point 81°C
Formula Weight 60.1g/mol
compliance-icons compliance-icons
Produktidentifikator
  • Isopropanol
Signalwort
  • Danger
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.