missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, Certified AR for Analysis, Fisher Chemical™

Product Code. 10497070 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic Container
Plastic-coated Glass Bottle
Unit Size:
1 L
10 L
2.5 L
25 L
5 L
500mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10497070 2.5 L Plastic-coated Glass Bottle 2.5 L
11453733 500 mL Plastic-coated Glass Bottle 500mL
10366430 1 L Amber Glass Bottle 1 L
10315720 2.5 L Amber Glass Bottle 2.5 L
10628143 2.5 L HDPE Bottle 2.5 L
10080320 5 L HDPE Bottle 5 L
10522992 10 L Plastic Container 10 L
10090320 25 L Metal Can 25 L
12656757 25 L Plastic Container 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10497070 Supplier Fisher Chemical Supplier No. P/7500/PB17
  • ONLINE EXCLUSIVE PRICE
    209.10 €
    174.00 € / 2.5 L
    Save 35.10 €
    16% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

C3H8O, CAS Number-67-63-0, 2-hydroxypropane, isopropanol, hartosol, 2-propanol, dimethylcarbinol, avantine, petrohol, sec-propyl alcohol, isopropyl alcohol, alkolave, 2.5L, 81 deg.C, CHEBI:17824, Colorless, 60.1g/mol, KFZMGEQAYNKOFK-UHFFFAOYSA-N, propan-2-ol, -89.5 deg.C, 11674, 60.096 | CAS: 67-63-0 | C3H8O | 60.096 g/mol

C3H8O, CAS Number-67-63-0, 2-hydroxypropane, isopropanol, hartosol, 2-propanol, dimethylcarbinol, avantine, petrohol, sec-propyl alcohol, isopropyl alcohol, alkolave, 2.5L, 81 deg.C, CHEBI:17824, Colorless, 60.1g/mol, KFZMGEQAYNKOFK-UHFFFAOYSA-N, propan-2-ol, -89.5 deg.C, 11674, 60.096

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number 11674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH 7
Melting Point -89.5°C
Color Colorless
Quantity 2.5 L
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Vapor Pressure 43mmHg at 20°C
Viscosity 2.27 mPaS at 20°C
Boiling Point 81°C
Formula Weight 60.1g/mol
Chemical Name or Material Isopropanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Propan-2-ol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.