missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, Molecular Biology Grade, Fisher BioReagents™

Product Code. 17140576 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Unit Size:
1 L
2.50 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17140576 1 L 1 L
17170576 500 mL 500mL
17150576 2.5 L 2.50 L
17160576 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17140576 Supplier Fisher BioReagents Supplier No. BP26181
  • ONLINE EXCLUSIVE PRICE
    151.00 €
    40.40 € / 1 L
    Save 110.60 €
    73% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For nucleic acid precipitation | CAS: 67-63-0 | C3H8O | 60.096 g/mol

For nucleic acid precipitation

Ultrapure reagent that can be used in fundamental applications such as purification and precipitation of nucleic acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

pH 7
Melting Point -89.5°C
Absorbance 0.015 AU max. at 254nm, 0.10 AU max. at 230nm, 0.20 AU max. at 220nm, 1.00 AU max. at 205nm
Color Colorless
Density 0.785g/cm³
Quantity 1 L
Packaging Amber Glass Bottle
Physical Form Liquid
Refractive Index 1.3740 to 1.3760 (at 25°C)
Residue after Evaporation 1ppm max.
Assay Percent Range ≥99.9 %
Linear Formula (CH3)2CHOH
Vapor Pressure 43mmHg at 20°C
Viscosity 2.27 mPa/s at 20°C
Water 0.05% max.
Boiling Point 81°C
Formula Weight 60.1
Percent Purity ≥99.9%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Isopropanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P378-In case of fire: Use to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2-Propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.