missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, 99+%, specified according to the requirements of Ph.Eur.

Product Code. 10326404 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
25 L
Unit Size:
1 L
2.5 L
25 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10326404 2.5 L 2.5 L
10441664 250 mL 250mL
10041174 1 L 1 L
15224724 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10326404 Supplier Thermo Scientific Acros Supplier No. 396980025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67-63-0 | C3H8O | 60.096 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -89.5°C
Absorbance 0.01 max. (EurPh) at 310nm, 0.02 max. (EurPh) at 290nm, 0.03 max. (EurPh) at 270nm, 0.1 max. (EurPh) at 250nm, 0.3 max. (EurPh) at 230nm
Color Brown to Purple
Appearance Clear colorless liquid
Density 0.7850g/mL
Quantity 2.5 L
Flash Point 12°C
Identification C: Passes Test (EP)
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Low Melting Solid
Refractive Index 1.3760 to 1.3780
Specific Gravity 0.785
Linear Formula (CH3)2CHOH
Viscosity 2.27 mPa.s (20°C)
Water 0.5% max.
Boiling Point 81.0°C to 83.0°C
Solubility Information Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, methylene chloride,, dmf and acetone, miscible with alcohol, ether and chloroform, insoluble in salts solutions
Formula Weight 60.1
Percent Purity ≥99%
Chemical Name or Material Isopropanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isopropanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.