missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 2-bromo-2-methylpropanoate, 97%, Thermo Scientific™

Product Code. 10420764
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10420764 100 g 100g
10160792 10 g 10g
10170792 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10420764 Supplier Thermo Scientific Maybridge Supplier No. SB01764FA
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 51368-55-9
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00055235
InChI Key UNZJYKKJZGIFCG-UHFFFAOYSA-N
Synonym isopropyl 2-bromo-2-methylpropanoate, isopropyl 2-bromo-2-methylpropionate, 2-bromo-2-methylpropionic acid isopropyl ester, isopropyl 2-bromoisobutyrate, propanoic acid, 2-bromo-2-methyl-, 1-methylethyl ester, isopropyl 2-bromo-2-methyl-propanoate, methylethyl 2-bromo-2-methylpropanoate, 2-bromo-iso-butyric acid isopropyl ester, propan-2-yl 2-bromanyl-2-methyl-propanoate, 2-bromoisobutyric acid isopropyl ester
PubChem CID 2796293
IUPAC Name propan-2-yl 2-bromo-2-methylpropanoate
SMILES CC(C)OC(=O)C(C)(C)Br

Specifications

CAS Min % 97.0
Density 1.2600g/mL
Assay Percent Range 97%
Linear Formula (CH3)2CBrCO2CH(CH3)2
Quantity 100 g
Formula Weight 209.08
Percent Purity 97%
Chemical Name or Material Isopropyl 2-bromo-2-methylpropanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.