missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 2-Bromo-2-methylpropanoate, TRC

Product Code. 30132817
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132817 5 g 5g
30006841 500 mg 500mg
30009692 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132817 Supplier Toronto Research Chemicals Supplier No. TRCI8746385G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51368-55-9
InChI Formula InChI=1S/C7H13BrO2/c1-5(2)10-6(9)7(3,4)8/h5H,1-4H3
IUPAC Name propan-2-yl 2-bromo-2-methylpropanoate
Molecular Formula C7H13O2Br
Molecular Weight (g/mol) 209.08
Chemical Name or Material Isopropyl 2-Bromo-2-methylpropanoate

Specifications

Quantity 5 g
Synonym 1-Methylethyl 2-Bromo-2-methylpropanoate, 2-Bromo-2-methylpropanoic Acid Isopropyl Ester, Isopropyl 2-Bromo-2-methylpropanoate, Isopropyl 2-Bromoisobutyrate, Isopropyl alpha-Bromoisobutyrate;
SMILES CC(C)OC(=O)C(C)(C)Br
Formula Weight 208.0099
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.