missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Acetoacetate, TRC

Product Code. 30396241
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30396241 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396241 Supplier Toronto Research Chemicals Supplier No. TRCI823725100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl Acetoacetate
CAS 542-08-5
Molecular Formula C7 H12 O3
Quantity 100 g
Synonym Butanoic acid, 3-oxo-, 1-methylethyl ester, Acetoacetic acid, isopropyl ester (6CI,8CI), 3-Oxobutanoic acid 1-methylethyl ester, 3-Oxobutanoic acid isopropyl ester, Isopropyl 3-oxobutanoate, Isopropyl 3-oxobutyrate, Isopropyl acetoacetate
InChI Formula InChI=1S/C7H12O3/c1-5(2)10-7(9)4-6(3)8/h5H,4H2,1-3H3
SMILES CC(C)OC(=O)CC(=O)C
IUPAC Name propan-2-yl 3-oxobutanoate
Molecular Weight (g/mol) 144.17
Formula Weight 144.0786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.