missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals

Product Code. 15763552 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15763552

Brand: Thermo Scientific J17886.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

  • Induces the lac operon in E. coli by binding and inactivating the lac repressor.
  • Selection for lac Y mutants.
  • Selection for recombinant plasmids containing β-galactosidase fusion proteins.
  • Induces the cellular content of lactose permease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Induces lac operon in E. coli by binding and inactivating lac repressor
  • Selection for lac Y mutants
  • Selection for recombinant plasmids containing beta-galactosidase fusion proteins
  • Induces cellular content of lactose permea
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-93-1
Molecular Formula C9H18O5S
Molecular Weight (g/mol) 238.298
MDL Number MFCD00063273
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
Synonym iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside
PubChem CID 656894
ChEBI CHEBI:61448
IUPAC Name (2 R,3 R,4 S,5 R,6 S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol
SMILES CC(C)SC1C(C(C(C(O1)CO)O)O)O

Specifications

Melting Point 109°C to 115°C
Packaging Plastic bottle
Quantity 1 g
Beilstein 4631
Formula Weight 238.3
Percent Purity ≥99%
Grade Ultrapure
Physical Form Powder
Chemical Name or Material Isopropyl-beta-D-thiogalactopyranoside (IPTG)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.