missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl bromoacetate, 97%

Product Code. 10269840 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10269840

Brand: Thermo Scientific Acros 412810250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29921-57-1 | C5H9BrO2 | 181.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29921-57-1
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00009886
InChI Key JCWLEWKPXYZHGQ-UHFFFAOYSA-N
Synonym isopropyl bromoacetate, acetic acid, bromo-, 1-methylethyl ester, acetic acid, 2-bromo-, 1-methylethyl ester, isopropyl 2-bromoacetate, brch2c o och ch3 2, bromoacetic acid isopropyl ester, i-propyl bromoacetate, acmc-1ciu8, isopropyl .alpha.-bromoacetate
PubChem CID 121593
IUPAC Name propan-2-yl 2-bromoacetate
SMILES CC(C)OC(=O)CBr

Specifications

Color Colorless to Yellow
Density 1.3990g/mL
Boiling Point 59.0°C to 61.0°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CO2CH(CH3)2
Refractive Index 1.4440 to 1.4460
Quantity 25 g
Specific Gravity 1.399
Formula Weight 181.03
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Isopropyl bromoacetate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Isopropyl bromoacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.