missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-β-D-thiogalactopyranoside, ≥99%, for biochemistry, contains ca. 15% 1,4-Dioxane, Thermo Scientific™

Product Code. 10431271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10431271 2.5 g Glass bottle 2.5g
10164660 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10431271 Supplier Thermo Scientific Acros Supplier No. 228560025
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 367-93-1 , 123-91-1
Molecular Formula C9H18O5S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00063273
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
Synonym iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside
PubChem CID 656894
ChEBI CHEBI:61448
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol
SMILES CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Color White
Loss on Drying 15% to 16% (90°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 2.5 g
Beilstein 17, IV, 3726
Merck Index 15, 5126
Specific Rotation Condition − 27.00 (20.00°C c=1,H2O)
Specific Rotation -25° to -33° (20°C, 589nm) (c=1, water)
Formula Weight 238.3
Percent Purity ≥99.0%
Grade Biochemical
Physical Form Crystalline Powder or Needles
Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.