missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-β-D-thiogalactopyranoside (IPTG) (Wht. Powd., Dioxane Crystalline), Fisher BioReagents™

Product Code. 10001273
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10001273 1 g Glass Bottle 1g
10715114 10 g Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10001273 Supplier Fisher BioReagents Supplier No. BP16201
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 367-93-1
Molecular Formula C9H18O5S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00063273
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 122°C
Color White
Quantity 1 g
Packaging Glass Bottle
Physical Form Solid
Merck Index 15, 5126
Formula Weight 238.3
Chemical Name or Material Isopropyl-β-D-thiogalactopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.