missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-β-D-thiogalactopyranoside, MP Biomedicals™

Product Code. 11487090 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
10 x 1 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487090 10 x 1 g 10g
11467090 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11487090 Supplier MP Biomedicals Supplier No. 11IPTG0010
 Restricted Item
Estimated Shipment: 04-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isopropyl-β-D-thiogalactopyranoside Genomic Grade, >99%, White Powder

  • IPTG is a galactose analogue not recognized by β-galactosidase. It used as a Lac operon inducer in bacteria.
  • At 1mM concentration, IPTG induces the β-galactosidase gene expression in bacteria, then it allows the selection of Lac+ recombinant bacteria in X-Gal containing medium.
  • It is used in conjunction with X-Gal or Bluo-Gal in blue-white selection of recombinant bacterial colonies that induce expression of the lac operon in Escherichia coli.

Chemical Identifiers

CAS 367-93-1
Molecular Formula C9H18O5S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00063273
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
Synonym IPTG, Isopropyl β-D-1-thiogalactopyranoside
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
SMILES CC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 110°C (literature)
Color White
pH 7
Quantity 10 x 1 g
Beilstein 4631
Solubility Information 10g/L in water.
Optical Rotation -28.9° to -32° (c=1, water)
Grade >99%
Physical Form Powder
compliance-icons
Product Identifier
  • Isopropyl-beta-D-thiogalactopyranoside (IPTG)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Isopropyl-lactopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.