missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl disulfide, 96%

Product Code. 10459040 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10459040 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10459040 Supplier Thermo Scientific Acros Supplier No. 122700250
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4253-89-8 | C6H14S2 | 150.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4253-89-8
Color Beige to White
Density 0.9430g/mL
Boiling Point 175°C to 176°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C6H14S2
Linear Formula [(CH3)2CH]2S2
Refractive Index 1.4900 to 1.492
MDL Number MFCD00008894
Quantity 25 g
Beilstein 01, 367
Specific Gravity 0.943
Synonym diisopropyl disulfide, isopropyl disulfide, diisopropyl disulphide, disulfide, bis 1-methylethyl, 2,2'-dithiodipropane, 1,2-diisopropyldisulfane, 2,5-dimethyl-3,4-dithiahexane, isopropyldisulfide, unii-bp550p623a, isopropyl disulfide 8ci
InChI Key LZAZXBXPKRULLB-UHFFFAOYSA-N
SMILES CC(C)SSC(C)C
IUPAC Name 2-(propan-2-yldisulfanyl)propane
Molecular Weight (g/mol) 150.3
PubChem CID 77932
Formula Weight 150.3
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Isopropyl disulfide, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isopropyl disulfide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.