missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl ether, 99+%, extra pure, stabilized with BHT

Product Code. 10501251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal drum
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10501251 2.5 L Glass bottle 2.5 L
10429110 1 L Glass bottle 1 L
10073113 25 L Metal drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10501251 Supplier Thermo Scientific Acros Supplier No. 180680025
  • ONLINE EXCLUSIVE PRICE
    65.95 €
    64.00 € / 2.5 L
    Save 1.95 €
    2% Off
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 108-20-3 | C6H14O | 102.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 108-20-3 , 128-37-0
MDL Number MFCD00008880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Spécification

Acidity 0.005% max.
Melting Point -85.5°C
Density 0.7200g/mL
Boiling Point 68°C
Flash Point -29°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C6H14O
Linear Formula [(CH3)2CH]2O
Refractive Index 1.3670 to 1.369
Quantity 2.5 L
UN Number 1159
Beilstein 01, 362
Residue after Evaporation 0.01% max.
Merck Index 15, 5258
Specific Gravity 0.72
Solubility Information Solubility in water: 9g/L (20°C). Other solubilities: miscible with most common organic solvents
Stabilizer 5 to 250ppm BHT
Molecular Weight (g/mol) 102.18
Viscosity 0.38 mPa.s (25°C)
Formula Weight 102.18
Vapor Pressure 180mbar at 20°C
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Isopropyl ether, Stabilized with BHT
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • Isopropyl ether
Mention d'avertissement
  • Danger
Catégorie de danger
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.