missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl ether, for analysis, stabilized

Product Code. 10380284 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10380284 1 L Glass bottle 1 L
10065253 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10380284 Supplier Thermo Scientific Acros Supplier No. 390660010
  • ONLINE EXCLUSIVE PRICE
    49.45 €
    48.00 € / 1 L
    Save 1.45 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 108-20-3 | C6H14O | 102.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-20-3 , 128-37-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00008880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Specifications

Acidity 0.0004 meq/g max.
Melting Point -85.5°C
Density 0.7200g/mL
Boiling Point 68°C
Flash Point -29°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula [(CH3)2CH]2O
Refractive Index 1.3670 to 1.369
Quantity 1 L
Merck Index 15, 5258
Specific Gravity 0.72
Solubility Information Solubility in water: 9g/L (20°C). Other solubilities: miscible with most common organic solvents
Stabilizer aprox. 50ppm BHT
Viscosity 0.38 mPa.s (25°C)
Formula Weight 102.18
Percent Purity ≥99.0%
Grade Analysis
Physical Form Liquid
Chemical Name or Material Isopropyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.