missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Ether (Stabilized with BHT), TRC

Product Code. 30389051
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389051 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389051 Supplier Toronto Research Chemicals Supplier No. TRCI824485100ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diisopropyl ether
CAS 108-20-3
Purity Grade Notes GC
Molecular Formula C6 H14 O
Quantity 100 mL
Synonym Isopropyl ether (8CI), 2,2'-Oxybis[propane], 2-Isopropoxypropane, Bis(isopropyl) ether, Di-1-methylethyl ether, Diisopropyl ether, Diisopropyl oxide
InChI Formula InChI=1S/C6H14O/c1-5(2)7-6(3)4/h5-6H,1-4H3
SMILES CC(C)OC(C)C
IUPAC Name 2-propan-2-yloxypropane
Molecular Weight (g/mol) 102.17
Formula Weight 102.1045
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.