missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl nitrate, 98%

Product Code. 15486718
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15486718 1 g 1g
15496718 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15486718 Supplier Thermo Scientific Alfa Aesar Supplier No. H60247.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1712-64-7 | C3H7NO3 | 105.093 g/mol

Isopropyl nitrate is used as a diesel cetane improver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl nitrate is used as a diesel cetane improver.

Solubility
Partly miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents, flammable substances, organic materials and metal powders.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1712-64-7
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00007041
InChI Key GAPFWGOSHOCNBM-UHFFFAOYSA-N
Synonym isopropyl nitrate, nitric acid, 1-methylethyl ester, 2-propyl nitrate, nitric acid, isopropyl ester, isopropylnitrate, isopropylester kyseliny dusicne, isopropylester kyseliny dusicne czech, ch3 2chono2, isonite, iso-propylnitrate
PubChem CID 15575
IUPAC Name propan-2-yl nitrate
SMILES CC(C)O[N+](=O)[O-]

Specifications

Density 1.04
Boiling Point 101°C to 102°C
Flash Point 12°C (54°F)
Linear Formula (CH3)2CHONO2
Refractive Index 1.391
Quantity 1 g
UN Number UN1222
Beilstein 1701405
Solubility Information Partly miscible in water.
Formula Weight 105.09
Percent Purity 98%
Chemical Name or Material Isopropyl nitrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.