missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Palmitate, TRC

Product Code. 30137769
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137769 50 g 50g
30138596 10 g 10g
30008453 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137769 Supplier Toronto Research Chemicals Supplier No. TRCI87271550G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-91-6
Molecular Formula C19 H38 O2
Molecular Weight (g/mol) 298.5
Synonym Isopropyl palmitate, Hexadecanoic acid, 1-methylethyl ester, 1-Methylethyl hexadecanoate, Palmitic acid, isopropyl ester (6CI,7CI,8CI), 1-Methylethyl hexadecanoate, Crodamol IPP, Deltyl, Deltyl Prime, Emcol IP, Emerest 2316, Estol 1517, Exceparl IPP, IPP, IPP-EX, Isopal, Isopalm, Isopropyl hexadecanoate, Isopropyl palmitate, Kessco IPP, Lexol IPP, NSC 69169, Neoderm IPP, Nikkol IPP, Nikkol IPP-EX, Palmsurf IPP 98, Propal, Sinnoester PIT, Stepan D 70, Stepan IPP, Versagel MP 1600, Wickenol 111
IUPAC Name propan-2-yl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC(C)C

Specifications

Quantity 50 g
InChI Formula InChI=1S/C19H38O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19(20)21-18(2)3/h18H,4-17H2,1-3H3
Formula Weight 298.2872
Chemical Name or Material Isopropyl hexadecanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.