missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl tetradecanoate, 98%

Product Code. 11479266
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479266 2500 mL 2500mL
11459266 100 mL 100mL
11469266 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479266 Supplier Thermo Scientific Alfa Aesar Supplier No. A18111.0F
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-27-0 | C17H34O2 | 270.457 g/mol

Isopropyl tetradecanoate is used as a solvent in perfume products and the oil phase in the preparation of formulations. It finds application in eye makeup, nail care products, soaps and detergents, hair care products, shaving products and other skin care products. It behaves as a non-aqueous component of the two-phase mouthwash, Dentyl pH, thereby it removes bacteria from the oral cavity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl tetradecanoate is used as a solvent in perfume products and the oil phase in the preparation of formulations. It finds application in eye makeup, nail care products, soaps and detergents, hair care products, shaving products and other skin care products. It behaves as a non-aqueous component of the two-phase mouthwash, Dentyl pH, thereby it removes bacteria from the oral cavity.

Solubility
Miscible with alcohol. Immiscible with water and glycerol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-27-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.457
MDL Number MFCD00008982
InChI Key AXISYYRBXTVTFY-UHFFFAOYSA-N
Synonym isopropyl myristate, isopropyl tetradecanoate, estergel, bisomel, isomyst, promyr, tetradecanoic acid, 1-methylethyl ester, deltyl extra, kesscomir, tegester
PubChem CID 8042
IUPAC Name propan-2-yl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC(C)C

Specifications

Density 0.85 g/mL
Boiling Point 191°C to 194°C (20 mmHg)
Flash Point 152°C (306°F)
Assay Percent Range 0.98
Quantity 2500 mL
Solubility Information Miscible with alcohol. Immiscible with water and glycerol.
Formula Weight 270.46
Physical Form Liquid
Chemical Name or Material Isopropyl Myristate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.