missing translation for 'onlineSavingsMsg'
Learn More

Isopropylamine, 99+%

Product Code. 11437307
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437307 2500 mL 2500mL
11417307 100 mL 100mL
11427307 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437307 Supplier Thermo Scientific Alfa Aesar Supplier No. A15044.0F
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-31-0 | C3H9N | 59.112 g/mol

Isopropylamine acts as a regulating agent for plastics and an additive in the petroleum industry. It is also useful as an intermediate for pharmaceuticals, coating materials, rubber chemicals and other organic products. It also finds application in binary chemical weapons in combination with isopropyl alcohol. Further, it is used as a solvent and to prepare other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropylamine acts as a regulating agent for plastics and an additive in the petroleum industry. It is also useful as an intermediate for pharmaceuticals, coating materials, rubber chemicals and other organic products. It also finds application in binary chemical weapons in combination with isopropyl alcohol. Further, it is used as a solvent and to prepare other chemicals.

Solubility
Miscible with water, chloroform, hexanes, acetone and ethanol.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and perchloryl fluoride.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-31-0
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.112
MDL Number MFCD00008082
InChI Key JJWLVOIRVHMVIS-UHFFFAOYSA-N
Synonym isopropylamine, 2-propanamine, 2-aminopropane, monoisopropylamine, 2-propylamine, sec-propylamine, 1-methylethylamine, isopropyl amine, isopropilamina, propane, 2-amino
PubChem CID 6363
ChEBI CHEBI:15739
IUPAC Name propan-2-amine
SMILES CC(C)N

Specifications

Melting Point -101°C
Density 0.691
Boiling Point 33°C to 34°C
Flash Point −32°C (−25°F)
Assay Percent Range ≥99%
Odor Amine-like
Linear Formula (CH3)2CHNH2
Refractive Index 1.375
Quantity 2500 mL
UN Number UN1221
Beilstein 605259
Sensitivity Air Sensitive
Merck Index 14,5209
Solubility Information Miscible with water,chloroform,hexanes,acetone and ethanol.
Formula Weight 59.11
Percent Purity ≥99%
Chemical Name or Material Isopropylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isopropylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.