missing translation for 'onlineSavingsMsg'
Learn More

Isopropylboronic acid pinacol ester, 97%

Product Code. 15496737
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496737 1 g 1g
15406747 5 g 5g
15416747 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. Vea la política de devoluciones
Código de producto. 15496737 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor H33861.03
 Restricted Item
Estimated Shipment: 17-11-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 76347-13-2 | C9H19BO2 | 170.059 g/mol

Isopropylboronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropylboronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 76347-13-2
Molecular Formula C9H19BO2
Molecular Weight (g/mol) 170.059
MDL Number MFCD05663856
InChI Key MECCSFDHAVAAAW-UHFFFAOYSA-N
Synonym 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropylboronic acid, pinacol ester, 2-isopropylboronic acid pinacol ester, isopropylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-methylethyl, 2-isopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 2-isoproyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-methylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 12727711
IUPAC Name 4,4,5,5-tetramethyl-2-propan-2-yl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C(C)C

Especificaciones

Boiling Point 115°C (60 mmHg)
Refractive Index 1.409
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 170.06
Percent Purity 98%
Chemical Name or Material Isopropylboronic acid pinacol ester

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.