missing translation for 'onlineSavingsMsg'
Learn More

Isopropylmagnesium bromide, 3M solution in 2-MeTHF, AcroSeal™

Product Code. 10557054 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10557054 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10557054 Supplier Thermo Scientific Acros Supplier No. 426781000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 920-39-8 | C3H7BrMg | 147.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 920-39-8, 96-47-9
Color Brown or Gray
Density 1.0800g/mL
Flash Point −11°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C3H7BrMg
Linear Formula (CH3)2CHMgBr
MDL Number MFCD00672110
Quantity 100 mL
UN Number 3399
Specific Gravity 1.08
Synonym isopropylmagnesium bromide, magnesium, bromo 1-methylethyl, isopropylmagnesiumbromide, bromo 1-methylethyl magnesium, iprmgbr, i-prmgbr, bromoisopropylmagnesium, i-propylmagnesiumbromide, i-propylmagnesium bromide, bromo-isopropyl-magnesium, grignard reagent
Solubility Information Solubility in water: reacts
InChI Key UCJIVFREPMUZDC-UHFFFAOYSA-M
SMILES CC(C)[Mg]Br
IUPAC Name magnesium;propane;bromide
Molecular Weight (g/mol) 147.30
PubChem CID 101909
Formula Weight 147.31
Physical Form Solution
Chemical Name or Material Isopropylmagnesium bromide, 3M solution in 2-Methyltetrahydrofuran
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isopropylmagnesium bromide, 3M solution in 2-Methyltetrahydrofuran
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • EUH019-May form explosive peroxides.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P233-Keep container tightly closed.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : Methyltetrahydrofuran, Bromo(1-methylethyl)magnesium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.