missing translation for 'onlineSavingsMsg'
Learn More

Isopropylmagnesium chloride, 1M in MeTHF

Product Code. 15408037
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408037 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15408037 Supplier Thermo Scientific Alfa Aesar Supplier No. H51155.AP
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1068-55-9 | C3H7ClMg | 102.84 g/mol

Isopropyl magnesium chloride is used primarily to add an iso-propyl group to carbonyl groups. This reagent can also be used as a strong base in aprotic solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl magnesium chloride is used primarily to add an iso-propyl group to carbonyl groups. This reagent can also be used as a strong base in aprotic solvents.

Solubility
It reacts violently with water.

Notes
Air & moisture sensitive. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1068-55-9
Molecular Formula C3H7ClMg
Molecular Weight (g/mol) 102.84
MDL Number MFCD00000466
InChI Key GPIBKUJXKCEZOH-UHFFFAOYSA-M
Synonym isopropylmagnesium chloride, magnesium, chloro 1-methylethyl, chloro 1-methylethyl magnesium, i-prmgcl, isopropylmagnesiumchloride, chloro isopropyl magnesium, isopropyl magnesium chloride, isopropylmagnesium chloride solution, iprmgcl, ipr-mgcl
PubChem CID 101955
IUPAC Name magnesium;propane;chloride
SMILES CC(C)[Mg]Cl

Specifications

Linear Formula (CH3)2CHMgCl
Quantity 500 mL
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information It reacts violently with water.
Formula Weight 102.85
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material Isopropylmagnesium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.