missing translation for 'onlineSavingsMsg'
Learn More

Isopropylmalonic acid, 98+%

Product Code. 11368275
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11368275

Brand: Thermo Scientific Alfa Aesar L10171.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 601-79-6 | C6H10O4 | 146.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 601-79-6
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD00021717
InChI Key DQEUFPARIOFOAI-UHFFFAOYSA-N
Synonym isopropylmalonic acid, 2-isopropylmalonic acid, malonic acid, isopropyl, 2-isopropylpropanedioic acid, propanedioic acid, 1-methylethyl, .alpha.-carboxyisovaleric acid, 2-propan-2-yl propanedioic acid, isopropylmalonsaure, isopropyl malonic acid, iso-propyl malonic acid
PubChem CID 69037
IUPAC Name 2-propan-2-ylpropanedioic acid
SMILES CC(C)C(C(O)=O)C(O)=O

Specifications

Melting Point 88°C to 90°C
Quantity 5 g
Beilstein 1723234
Formula Weight 146.14
Percent Purity ≥98%
Chemical Name or Material Isopropylmalonic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.