missing translation for 'onlineSavingsMsg'
Learn More

Isoproterenol Hydrochloride, TRC

Product Code. 30408609
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30408609 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408609 Supplier Toronto Research Chemicals Supplier No. TRCI874200100MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isoprenaline Hydrochloride
CAS 51-30-9
Molecular Formula C11 H17 N O3 . Cl H
Quantity 100 mg
Synonym Isoprenaline hydrochloride, (1RS)-1-(3,4-Dihydroxyphenyl)-2-[(1-methylethyl)amino]ethanol hydrochloride, 1,2-Benzenediol, 4-[1-hydroxy-2-[(1-methylethyl)amino]ethyl]-, hydrochloride (1:1), 1,2-Benzenediol, 4-[1-hydroxy-2-[(1-methylethyl)amino]ethyl]-, hydrochloride (9CI), Benzyl alcohol, 3,4-dihydroxy-α-[(isopropylamino)methyl]-, hydrochloride (8CI), Protocatechuyl alcohol, α-(isopropylaminomethyl)-, hydrochloride (5CI), (±)-Isoprenaline hydrochloride, (±)-Isopropylnoradrenaline hydrochloride, (±)-Isoproterenol hydrochloride, 1-(3,4-Dihydroxyphenyl)-2-(isopropylamino)ethanol hydrochloride, 3,4-Dihydroxy-α-(isopropylaminomethyl)benzyl alcohol hydrochloride, Aerolone, Aerotrol, Asthpul, DL-Isoprenaline hydrochloride, DL-Isoproterenol hydrochloride, Euspiran, Isadrine, Isomenyl, Isoprel, Isoprenaline chloride, Isopropylarterenol hydrochloride, Isoproterenol hydrochloride, Isovon, Isuprel, Mistarel, NSC 37745, NSC 89747, Proternol, Saventrine, Suscardia, Vapo-Iso, dl-Isadrine hydrochloride, dl-Isoprenaline hydrochloride, dl-Isopropylnoradrenaline hydrochloride, dl-Isopropylnorepinephrine hydrochloride, dl-Isoproterenol hydrochloride, α-(Isopropylaminomethyl)-3,4-dihydroxybenzyl alcohol hydrochloride
InChI Formula InChI=1S/C11H17NO3.ClH/c1-7(2)12-6-11(15)8-3-4-9(13)10(14)5-8;/h3-5,7,11-15H,6H2,1-2H3;1H
SMILES Cl.CC(C)NCC(O)c1ccc(O)c(O)c1
IUPAC Name 4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;hydrochloride
Molecular Weight (g/mol) 247.72
Formula Weight 247.0975
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.