missing translation for 'onlineSavingsMsg'
Learn More

Isopulegol, mixture of isomers, 97%

Product Code. 11469746
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469746 50 g 50g
11459746 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469746 Supplier Thermo Scientific Alfa Aesar Supplier No. B20260.18
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7786-67-6 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7786-67-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00134655
InChI Key ZYTMANIQRDEHIO-UHFFFAOYNA-N
Synonym p-menth-8-en-3-ol, isopulegol, 8 9-p-menthen-3-ol, 5-methyl-2-prop-1-en-2-yl cyclohexanol, cyclohexanol, 5-methyl-2-1-methylethenyl, 1-methyl-4-isopropenylcyclohexan-3-ol, dl-isopulegol, p-8 9-menthen-3-ol, 5-methyl-2-1-methylvinyl cyclohexan-1-ol, 5-methyl-2-prop-1-en-2-yl cyclohexan-1-ol
PubChem CID 24585
IUPAC Name 5-methyl-2-prop-1-en-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(O)C1)C(C)=C

Specifications

Density 0.91
Boiling Point 90°C to 92°C (12 mmHg)
Flash Point 78°C (172°F)
Refractive Index 1.4725
Quantity 50 g
Formula Weight 154.25
Percent Purity 97%
Chemical Name or Material Isopulegol, mixture of isomers
compliance-icons
Product Identifier
  • Isopulegol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Isopulegol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.