missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-4-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10706905
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10706905 1 g Amber Glass Bottle 1g
10451684 250 mg Amber Glass Bottle 250mg
2 options
This item is not returnable. View return policy

Product Code. 10706905

Brand: Thermo Scientific Maybridge CC04904DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 22960-16-3
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD00829440
InChI Key RNQQJLYJLDQGGL-UHFFFAOYSA-N
Synonym isoquinoline-4-carboxaldehyde, 4-isoquinolinecarboxaldehyde, 4-formylisoquinoline, pubchem15962, acmc-20aig4, 4-isoquinolinecarbaldehyde, ksc201k4f, 4-isoquinolinecarboxaldehyde 7ci,8ci,9ci
PubChem CID 10868870
IUPAC Name isoquinoline-4-carbaldehyde
SMILES C1=CC=C2C(=C1)C=NC=C2C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 157.17
Percent Purity 97%
Chemical Name or Material Isoquinoline-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.