missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10796895
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10796895 250 mg Amber Glass Bottle 250mg
10481494 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10796895 Supplier Thermo Scientific Maybridge Supplier No. CC04901CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 7159-36-6
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00094344
InChI Key MCVMLYSLPCECGO-UHFFFAOYSA-N
Synonym 4-isoquinolinecarboxylic acid, 4-carboxyisoquinoline, 4-isoquinolinecarboxylicacid, isoquinoline-4-carboxylicacid, 4-carboxy-isoquinoline, acmc-209oja, isoquinoline-4-carboxylic, isoquinolin-4-carboxylic acid, n-2-aminoethyl pyrrolidine; 2-1-pyrrolidino ethylamine
PubChem CID 459768
IUPAC Name isoquinoline-4-carboxylic acid
SMILES OC(=O)C1=CN=CC2=CC=CC=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 173.17
Percent Purity 97%
Chemical Name or Material Isoquinoline-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.