missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-5-carboxaldehyde, 97%

Product Code. 11315435
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11315435 250 mg 250mg
11305435 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11315435 Supplier Thermo Scientific Alfa Aesar Supplier No. L19636.MD
 Restricted Item
Estimated Shipment: 29-12-2026
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 80278-67-7 | C10H7NO | 157.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 80278-67-7
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD03412483
InChI Key ILRSABOCKMOFGW-UHFFFAOYSA-N
Synonym 5-isoquinolinecarboxaldehyde, isoquinoline-5-carboxaldehyde, 5-formylisoquinoline, ksc447q7n, 5-isoquinolinecarbaldehyde, 5-isoquinolinecarboxaldehyde 6ci,9ci
PubChem CID 7016853
IUPAC Name isoquinoline-5-carbaldehyde
SMILES C1=CC2=C(C=CN=C2)C(=C1)C=O

Specifikationer

Melting Point 113°C to 116°C
Quantity 250 mg
Beilstein 112875
Sensitivity Air Sensitive
Formula Weight 157.17
Percent Purity 97%
Chemical Name or Material Isoquinoline-5-carboxaldehyde

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.