missing translation for 'onlineSavingsMsg'
Learn More

Isosorbide 5-Mononitrate, TRC

Product Code. 30137076
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137076 1 g 1g
30011562 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137076 Supplier Toronto Research Chemicals Supplier No. TRCI9021001G
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16051-77-7
Molecular Formula C6 H9 N O6
Molecular Weight (g/mol) 191.14
Synonym Isosorbide Mononitrate, Isosorbide Dinitrate Imp. C (EP), Isosorbide 5-Nitrate, 1,4:3,6-Dianhydro-D-glucitol 5-nitrate, D-1,4:3,6-Dianhydro-D-glucitol 5-nitrate, Furo[3,2-b]furan, D-glucitol deriv., AHR 4698, BM 22-145, Conpin, Conpin Retardkaps, Corangin, Corangin SR, Dilavenil, Duride, Elan, Elantan, Elantan Long, Elantan Retard, Geomatrix, IS 5MN, ISMN, Imdur, Imdur 60, Imdur Durules, Imodur, Imtrate, Ismexin, Ismo, Ismo 20, Ismox, Isomon, Isomonat, Isomonit, Isopen 20, Isosorbide 5-mononitrate, Isosorbide 5-nitrate, Isosorbide mononitrate, Iturol, Medocor, Momo Mack, Monicor, Monis, Monit, Monit 20, Monizid, Mono Corax, Mono Corax Retard, Mono-Cedocard, Mono-Mack, Mono-Sanorania, Monoclair, Monocord 20, Monocord 40, Monocord 50SR, Monodur Durules, Monoket, Monoket OD, Monoket Retard, Monolong, Monolong 40, Monolong 60, Mononit, Mononit 20, Mononit 40, Mononit Retard 50, Monopront, Monosorb, Monosorbitrate, Monosordil, Monotrate, Monovas, Multitab, Nitramin, Olicard, Ormox, Pentacard, Pentacard 20, Vasotrate
IUPAC Name [(3S,3aR,6R,6aS)-3-hydroxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-6-yl] nitrate
SMILES O[C@H]1CO[C@@H]2[C@@H](CO[C@H]12)O[N+](=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H9NO6/c8-3-1-11-6-4(13-7(9)10)2-12-5(3)6/h3-6,8H,1-2H2/t3-,4+,5+,6+/m0/s1
Formula Weight 191.043
Chemical Name or Material Isosorbide Mononitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.