missing translation for 'onlineSavingsMsg'
Learn More

Isosorbide, 98%

Product Code. 10103401 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10103401 5 g Glass bottle 5g
10153491 100 g Glass bottle 100g
10096343 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10103401 Supplier Thermo Scientific Acros Supplier No. 351910050
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 652-67-5 | C6H10O4 | 146.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652-67-5
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD00064827
InChI Key KLDXJTOLSGUMSJ-JGWLITMVSA-N
Synonym isosorbide, isobide, devicoran, hydronol, ismotic, sorbid, +-d-isosorbide, vascardin dinitrate, 3r,3ar,6s,6ar-hexahydrofuro 3,2-b furan-3,6-diol, 1,4-dianhydrosorbitol
PubChem CID 12597
IUPAC Name (3S,3aR,6R,6aR)-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-3,6-diol
SMILES C1C(C2C(O1)C(CO2)O)O

Specifications

Melting Point 56.0°C to 62.0°C
Color Beige or White to Yellow
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystals, Crystalline Solid or Mass
Assay Percent Range 97.5% min. (GC)
Merck Index 15, 5270
Formula Weight 146.14
Percent Purity 98%
Chemical Name or Material Isosorbide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.