missing translation for 'onlineSavingsMsg'
Learn More

Isosorbide mononitrate, MedChemExpress

Product Code. 16492249
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492249 1 g 1g
16491028 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492249 Supplier Medchem Express Supplier No. HYB0642
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Isosorbide mononitrate(Isosorbide-5-mononitrate) is a nitrate-class compound used for angina pectoris; acts by dilating the blood vessels so as to reduce the blood pressure.

Isosorbide mononitrate(Isosorbide-5-mononitrate) is a nitrate-class compound used for angina pectoris

Specifications

Chemical Name or Material Isosorbide mononitrate
CAS 16051-77-7
Color White
Purity Grade Notes Research
Molecular Formula C6H9NO6
Quantity 1 g
Synonym Isosorbide-5-mononitrate
Solubility Information DMSO : 100 mg/mL (523.18 mM; Need ultrasonic) ∣H2O : 50 mg/mL (261.59 mM; Need ultrasonic)
SMILES O=[N+]([O-])O[C@H]1[C@](OC[C@@H]2O)([H])[C@]2([H])OC1
Molecular Weight (g/mol) 191.14
Formula Weight 191.14
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.