missing translation for 'onlineSavingsMsg'
Learn More

Isosorbide, TRC

Product Code. 30135487
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135487 1 g 1g
30138719 10 g 10g
30010073 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135487 Supplier Toronto Research Chemicals Supplier No. TRCI9016801G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652-67-5
Molecular Formula C6 H10 O4
Molecular Weight (g/mol) 146.14
Synonym Isosorbide, D-Glucitol, 1,4:3,6-dianhydro-, Glucitol, 1,4:3,6-dianhydro-, D- (8CI), Sorbitol, 1,4:3,6-dianhydro- (6CI), Furo[3,2-b]furan, D-glucitol deriv., (+)-D-Isosorbide, 1,4:3,6-Dianhydro-D-glucitol, 1,4:3,6-Dianhydro-D-sorbitol, 1,4:3,6-Dianhydrosorbitol, AT 101, Devicoran, Hydronol, Ismotic, Isobide, NSC 40725
IUPAC Name (3S,3aR,6R,6aR)-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-3,6-diol
SMILES O[C@@H]1CO[C@@H]2[C@@H](O)CO[C@H]12

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H10O4/c7-3-1-9-6-4(8)2-10-5(3)6/h3-8H,1-2H2/t3-,4+,5-,6-/m1/s1
Formula Weight 146.0579
Chemical Name or Material Isosorbide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.