missing translation for 'onlineSavingsMsg'
Learn More

Isothiazol-3(2H)-one, TRC

Product Code. 30390130
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30390130 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390130 Supplier Toronto Research Chemicals Supplier No. TRCI917620100MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Isothiazol-3(2H)-one
CAS 1003-07-2
Molecular Formula C3 H3 N O S
Quantity 100 mg
Synonym 4-Isothiazolin-3-one (7CI,8CI), 1,2-Isothiazole-3-one, 1,2-Thiazol-3-one, 3-Hydroxyisothiazole, 3-Isothiazolol, 3-Isothiazolone, 3-Oxo-2,3-dihydroisothiazole, A 01, Duo Mei Jing, Ebotec aqua, GT 2060, GY 506, Isothiazolin-3-one, Isothiazolinone, NSC 93489, PT 490, Xarcin AB
InChI Formula InChI=1S/C3H3NOS/c5-3-1-2-6-4-3/h1-2H,(H,4,5)
SMILES O=C1NSC=C1
IUPAC Name 1,2-thiazol-3-one
Molecular Weight (g/mol) 101.127
Formula Weight 100.9935
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.