missing translation for 'onlineSavingsMsg'
Learn More

Isovaleronitrile, 98%

Product Code. 10031034 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Product Code. Quantity Packaging unitSize
10031034 25 g Glass bottle 25g
10692255 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10031034

Brand: Thermo Scientific Acros 427560250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 625-28-5 | C5H9N | 83.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-28-5
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00001944
InChI Key QHDRKFYEGYYIIK-UHFFFAOYSA-N
Synonym isovaleronitrile, butanenitrile, 3-methyl, isoamylnitrile, isobutyl cyanide, isopentane nitrile, 3-methylbutyronitrile, butyronitrile, 3-methyl, 2-methylbutane secondary mononitrile, unii-n2g72x091l, isopentanenitrile
PubChem CID 12244
IUPAC Name 3-methylbutanenitrile
SMILES CC(C)CC#N

Specifications

Color Colorless
Density 0.7950g/mL
Boiling Point 128.0°C to 130.0°C (730.0 mmHg)
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CN
Quantity 25 g
Specific Gravity 0.795
Formula Weight 83.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isovaleronitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isovaleronitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.