missing translation for 'onlineSavingsMsg'
Learn More

Isovanillin, ≥99.5% (HPLC), Honeywell™ Fluka™

Product Code. 15615900 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Poly bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15615900 1 g Poly bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15615900 Supplier Honeywell-Fluka Supplier No. 599271G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Matrix substance for MALDI-MS,≥99.5% (HPLC)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 621-59-0
Melting Point 113°C to 116°C
Color Brown
Boiling Point 179°C
Packaging Poly bottle
Molecular Formula C8H8O3
MDL Number MFCD00003369
Quantity 1 g
UN Number NONH for all modes of transport
Beilstein 1073021
Synonym isovanillin, isovanilline, 3-hydroxy-p-anisaldehyde, 5-formylguaiacol, benzaldehyde, 3-hydroxy-4-methoxy, iso-vanillin, 3-hydroxyanisaldehyde, isovanilin, isovanicaline, 3-hydroxy-4-methoxy-benzaldehyde
InChI Key JVTZFYYHCGSXJV-UHFFFAOYSA-N
SMILES COC1=C(C=C(C=C1)C=O)O
IUPAC Name 3-hydroxy-4-methoxybenzaldehyde
Molecular Weight (g/mol) 152.149
PubChem CID 12127
Formula Weight 152.15g/mol
Percent Purity ≥99.5% (HPLC)
Physical Form Crystalline
Chemical Name or Material Isovanillin, Matrix substance for MALDI-MS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.