missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole-3-carboxylic acid, 97%

Product Code. 15444728
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15444728 1 g 1g
15454728 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444728 Supplier Thermo Scientific Alfa Aesar Supplier No. H58438.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3209-71-0 | C4H3NO3 | 113.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3209-71-0
Molecular Formula C4H3NO3
Molecular Weight (g/mol) 113.07
MDL Number MFCD06738796
InChI Key UXYRXGFUANQKTA-UHFFFAOYSA-N
Synonym 3-isoxazolecarboxylic acid, isoxazole-3-carboxylic acid, isoxazole-3-carboxylicacid, 3-isoxazolecarboxylicacid, 3-carboxyisoxazole, isoxazole-3-carboxylic, pubchem12341, acmc-209hqw, ksc222a3r, aronis24546
PubChem CID 11286453
IUPAC Name 1,2-oxazole-3-carboxylic acid
SMILES OC(=O)C1=NOC=C1

Specifications

Quantity 1 g
Beilstein 110919
Formula Weight 113.07
Percent Purity 97%
Chemical Name or Material Isoxazole-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.