missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole-5-carbonyl chloride, TECH, Thermo Scientific™

Product Code. 10702293
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10702293 1 g 1g
10106963 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10702293 Supplier Thermo Scientific Maybridge Supplier No. CC10302DA
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 62348-13-4
Molecular Formula C4H2ClNO2
Molecular Weight (g/mol) 131.52
MDL Number MFCD00067868
InChI Key NASLINFISOTVJJ-UHFFFAOYSA-N
Synonym isoxazole-5-carbonyl chloride, 5-isoxazolecarbonyl chloride, isoxazole-5-carbonylchloride, isoxazole-5-carboxylic acid chloride, acmc-1bhwq, 5-isoxazolecarbonylchloride, isoxazol-5-carbonyl chloride, isoxazole-5-carbonyl-chloride, 5-isoxazolecarbonyl chloride 9ci
PubChem CID 2736707
IUPAC Name 1,2-oxazole-5-carbonyl chloride
SMILES ClC(=O)C1=CC=NO1

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 131.52
Percent Purity 90%
Chemical Name or Material Isoxazole-5-carbonyl chloride
compliance-icons
Product Identifier
  • Isoxazole-5-carbonyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.