missing translation for 'onlineSavingsMsg'
Learn More

Isoxazole, 99%

Product Code. 15438799
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438799 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15438799 Supplier Thermo Scientific Alfa Aesar Supplier No. L01607.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 288-14-2 | C3H3NO | 69.063 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 288-14-2
Density 1.078
Boiling Point 93°C to 95°C
Flash Point 8°C (46°F)
Molecular Formula C3H3NO
Refractive Index 1.427
MDL Number MFCD00003149
Quantity 5 g
UN Number UN1993
Beilstein 103773
Synonym isoxazole, 1-oxa-2-azacyclopentadiene, isooxazole, unii-00srw0m6pw, 00srw0m6pw, isoxazoles, pubchem8623, acmc-209h4x, ksc204o6n
InChI Key CTAPFRYPJLPFDF-UHFFFAOYSA-N
SMILES C1=CON=C1
IUPAC Name 1,2-oxazole
Molecular Weight (g/mol) 69.063
PubChem CID 9254
ChEBI CHEBI:35595
Formula Weight 69.06
Percent Purity 99%
Chemical Name or Material Isoxazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.