missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Isradipine

Product Code. 30001695
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001695 10 mg 10mg
18003802 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001695 Supplier R&D Systems™ Supplier No. 2004/10
  • ONLINE EXCLUSIVE PRICE
    227.13 €
    203.70 € / 10mg
    Save 23.43 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. CaV1.x blocker

Potent and selective L-type voltage-gated Ca2+ channel blocker. EC50 and pA2 values are 1.4 nM and 10.3 for relaxation of depolarization- and Ca2+-induced contractions of rabbit aorta respectively; EC25 = 0.45 nM for reduction in rate of spontaneously beating guinea pig right atria. Long-acting antihypertensive agent in vivo, exerting primary effects on vascular tissue with secondary negative chronotropic action.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75695-93-1
Molecular Formula C19H21N3O5

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 371.39
Purity >99%
Molecular Weight (g/mol) 371.39
Chemical Name or Material Isradipine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.