missing translation for 'onlineSavingsMsg'
Learn More

Isradipine, MedChemExpress

Product Code. 16494307
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494307 50 mg 50mg
16468428 10 mg 10mg
16428186 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16494307 Supplier Medchem Express Supplier No. HYB0233
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Isradipine (PN 200-110) is an orally active L-type calcium channel blocker. Isradipine, as a powerful peripheral vasodilator, is a dihydropyridine calcium antagonist with selective actions on the heart as well as the peripheral circulation. Isradipine is a potentially viable neuroprotective agent for Parkinson's disease.

Isradipine (PN 200-110) is an orally active L-type calcium channel blocker. Isradipine, as a powerful peripheral vasodilator, is a dihydropyridine calcium antagonist with selective actions on the heart as well as the peripheral circulation. Isradipine is a potentially viable neuroprotective agent for Parkinson's disease.

Specifications

Chemical Name or Material Isradipine
CAS 75695-93-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H21N3O5
Quantity 50 mg
Synonym PN 200-110
Solubility Information DMSO : ≥ 100 mg/mL (269.26 mM)
SMILES O=C(C1=C(C)NC(C)=C(C(OC(C)C)=O)C1C2=CC=CC3=NON=C23)OC
Molecular Weight (g/mol) 371.39
Formula Weight 371.39
Percent Purity 99.69%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.