missing translation for 'onlineSavingsMsg'
Learn More

Itaconic acid, 99+%

Product Code. 10457700 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10457700 250 g Plastic bottle 250g
10549040 1 kg Plastic bottle 1kg
11945031 5 kg Plastic bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10457700 Supplier Thermo Scientific Acros Supplier No. 122812500

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 97-65-4 | C5H6O4 | 130.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-65-4
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.1
InChI Key LVHBHZANLOWSRM-UHFFFAOYSA-N
Synonym itaconic acid, 2-methylenesuccinic acid, methylenesuccinic acid, methylenebutanedioic acid, propylenedicarboxylic acid, itaconate, 2-propene-1,2-dicarboxylic acid, butanedioic acid, methylene, succinic acid, methylene, 2-methylenebutanedioic acid
PubChem CID 811
ChEBI CHEBI:30838
IUPAC Name 2-methylidenebutanedioic acid
SMILES C=C(CC(=O)O)C(=O)O

Specifications

Melting Point 165.0°C to 168.0°C
Color Beige to White
Density 1.5700g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula HO2CCH2C(=CH2)CO2H
Quantity 250 g
Beilstein 02,76
Merck Index 15,529
Specific Gravity 1.57
Solubility Information Solubility in water: 95g/L (25°C). Other solubilities: soluble in alcohol and acetone,slightly soluble in most other common organic,solvents
Formula Weight 130.1
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Itaconic acid
Show More Show Less
compliance-icons
Product Identifier
  • Itaconic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: butanedioic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.