missing translation for 'onlineSavingsMsg'
Learn More

Itaconic acid, 99%

Product Code. 11453027
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453027 250 g 250g
11443027 1000 g 1000g
11463027 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453027 Supplier Thermo Scientific Alfa Aesar Supplier No. A15566.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 97-65-4 | C5H6O4 | 130.099 g/mol

Itaconic acid is used in the preparation of acrylonitrile-butadiene-styrene and acrylate latexes. It is also used to prepare poly-itaconic acid, resins biofuel components and ionomer cements. It finds application in the textile, chemical and pharmaceutical industries. It is also used as an additive in fibers and ion exchange resins to increase abrasion, waterproofing, physical resistance, dying affinity and better duration. Further, it acts as a co-monomer used in the preparation of acrylic fibers and rubbers, reinforced glass fiber, artificial diamonds and lens. In addition to this, it acts as a binder and sizing agent in non-weaving fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Itaconic acid is used in the preparation of acrylonitrile-butadiene-styrene and acrylate latexes. It is also used to prepare poly-itaconic acid, resins biofuel components and ionomer cements. It finds application in the textile, chemical and pharmaceutical industries. It is also used as an additive in fibers and ion exchange resins to increase abrasion, waterproofing, physical resistance, dying affinity and better duration. Further, it acts as a co-monomer used in the preparation of acrylic fibers and rubbers, reinforced glass fiber, artificial diamonds and lens. In addition to this, it acts as a binder and sizing agent in non-weaving fibers.

Solubility
Soluble in water, acetone, methanol, hexane and ethanol. Slightly soluble in benzene, chloroform, carbon disulfide and petroleum ether.

Notes
Hygroscopic. Moisture sensitive. Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-65-4
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.099
MDL Number MFCD00004260
InChI Key LVHBHZANLOWSRM-UHFFFAOYSA-N
Synonym itaconic acid, 2-methylenesuccinic acid, methylenesuccinic acid, methylenebutanedioic acid, propylenedicarboxylic acid, itaconate, 2-propene-1,2-dicarboxylic acid, butanedioic acid, methylene, succinic acid, methylene, 2-methylenebutanedioic acid
PubChem CID 811
ChEBI CHEBI:30838
IUPAC Name 2-methylidenebutanedioic acid
SMILES C=C(CC(=O)O)C(=O)O

Specifications

Melting Point 165°C to 169°C
Density 1.63
Boiling Point 268°C
Quantity 250 g
Beilstein 1759501
Sensitivity Hygroscopic
Merck Index 14,5242
Solubility Information Soluble in water,acetone,methanol,hexane and ethanol. Slightly soluble in benzene,chloroform,carbon disulfide and petroleum ether.
Formula Weight 130.1
Percent Purity 99%
Chemical Name or Material Itaconic acid
Show More Show Less
compliance-icons
Product Identifier
  • Itaconic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Itaconic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.